3-Methoxythiophenol 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-Methoxybenzenethiol, 1-Methoxy-3-mercaptobenzene, 3-Mercaptoanisol, 3-Methoxybenzene-1-thiol, 3-Methoxyphenyl mercaptan, m-Methoxythiophenol
A10648-03
A10648-06
A10648-14
CAAAAA10648-03EA
42.82
CAD
CAAAAA10648-03
CAAAA10648-06
CAAAA10648-14
3-Methoxythiophenol 98%
3-Methoxythiophenol
Purity: 98%
CAS No.: 15570-12-4
Molecular Formula: C7H8OS
Formula Weight: 140.21
Flash Point: 96°(204°F)
Refractive Index: 1.5865
Odor: Unpleasant
Boiling Point: 225-227°
Beilstein Reference: 2041496
MDL No.: MFCD00004841
Formula:
C₇H₈OS MW: 140.21 g/mol Boiling Pt: 223 Density: 1.136 Flash Pt: 96 °C (204 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004841 CAS Number: 15570-12-4 EINECS: 239-617-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...