Manganese(IV) oxide ≥99.996% (metals basis), Puratronic®

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Manganese dioxide, Manganese(IV) oxide, activated, Manganese peroxide, Mangan(IV)-oxid

10805-06 10805-14 10805-22
CAAA10805-06EA 95.91 CAD
CAAA10805-06 CAAA10805-14 CAAA10805-22
Manganese(IV) oxide ≥99.996% (metals basis), Puratronic®
Manganese (IV) oxide

Powder. Purity based on metal contaminates only.


Purity: 99.996% CAS No.: 1313-13-9 Molecular Formula: MnO2 Formula Weight: 86.94 Physical Form: Powder Density: 5.026 Appearance: Black Melting Point: 535° dec. Merck Reference: 14,5730 MDL No.: MFCD00003463

Formula: MnO₂
MW: 86.94 g/mol
Melting Pt: 535 °C (dec.)
Density: 5.026
Storage Temperature: Ambient
MDL Number: MFCD00003463
CAS Number: 1313-13-9
EINECS: 215-202-6
Merck Index: 14,5730

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR