Lignoceric acid ≥96.0% (by titrimetric analysis)
Supplier: TCI America
|
Warning
|
T0076-1G
T0076-25G
T0076-5G
TCT0076-001GEA
154.84
CAD
TCT0076-001G
TCT0076-025G
TCT0076-5G
Lignoceric acid ≥96.0% (by titrimetric analysis)
Lignoceric acid
CAS Number: 557-59-5
MDL Number: MFCD00002810
Molecular Formula: C24H48O2
Molecular Weight: 368.65
Purity/Analysis Method: >96.0% (T)
Form: Crystal
Color: White
Melting point (°C): 84
Product Categories: Bioscience, Lipids, Higher Fatty Acids & Higher Alcohols
Other Product Names / Synonyms: Tetracosanoic Acid
Formula:
C₂₄H₄₈O₂ Boiling Pt: 406 °C (1013 hPa) Melting Pt: 84 °C Density: 0.8207 g/cm³ (100 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002810 CAS Number: 557-59-5 EINECS: 209-180-7 Merck Index: 13,05509 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...