Linoleic acid ≥97.0%

Supplier: TCI America

Synonyms: (9Z,12Z)-Octadeca-9,12-dienoic acid, cis,cis-9,12-Octadecadienoic acid, 9Z,12Z-Octadecadienoic acid

L0124-5G L0124-25G
TCL0124-005GEA 120.59 CAD
TCL0124-005G TCL0124-025G
Linoleic acid ≥97.0%
Linoleic acid

CAS Number: 60-33-3
MDL Number: MFCD00064241
Molecular Formula: C18H32O2
Molecular Weight: 280.45
Purity/Analysis Method: >97.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 177
Melting point (°C): -5
Flash Point (°C): 113
Specific Gravity (20/20): 0.90
Lambda max.: 270 nm (H2O)Storage Temperature: 0-10°C


Product Categories: Bioscience, Lipids, Higher Fatty Acids & Higher Alcohols, Antioxidants
Other Product Names / Synonyms: 9,12-Octadecadienoic Acid

Formula: C₁₈H₃₂O₂
MW: 280.45 g/mol
Boiling Pt: 360
Melting Pt: -5
Density: 0.90 (20 °C)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00064241
CAS Number: 60-33-3
EINECS: 200-470-9
Merck Index: 13,05527
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bf291b412effb9342c01769ad8e0b142940020f8&vid=bf291b412effb9342c01769ad8e0b142940020f8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR