Round-Bottom Boiling Flasks with Two Necks, Heavy Wall, Chemglass
Supplier: Chemglass
CG-1520-16 CG-1520-18 CG-1520-20 CG-1520-49 CG-1520-52 CG-1520-01 CG-1520-02 CG-1520-03 CG-1520-04 CG-1520-06 CG-1520-09 CG-1520-10 CG-1520-12 CG-1520-14 CG-1520-41 CG-1520-43 CG-1520-44 CG-1520-45 CG-1520-46 CG-1520-47 CG-1520-48 CG-1520-50 CG-1520-51 CG-1520-54 CG-1520-55 CG-1520-57
80069-158EA 443.4 CAD
80069-158 80069-160 80069-162 80069-190 80069-196 80069-132 80069-134 80069-136 80069-138 80069-142 80069-144 80069-146 80069-150 80069-154 80069-174 80069-176 80069-178 80069-180 80069-182 80069-184 80069-186 80069-188 80069-192 80069-194 80069-200 80069-202 80069-206
Round-Bottom Boiling Flasks with Two Necks, Heavy Wall, Chemglass
Flasks Round Bottom Flasks
Round bottom, heavy wall, two-neck flask with [ST] outer joints on both the center neck and 20°-angled side neck
Flasks with 10/30 side neck are for use with non mercury joint thermometers and to aid in clamping of the flask, the 29/26 center neck flasks have a groove tooled just below the grind.
Both side neck and center neck have the same joint size.
Ordering information: Flasks with joint sizes other than those listed are available.