Lithium carbonate 99,0, powder, AR® ACS, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning


M.O.C. Category N

MACR5840-02 MACR5840-06
CAMK584002CS 3428.62 CAD
CAMK584002 CAMK5840-06
Lithium carbonate 99,0, powder, AR® ACS, Macron Fine Chemicals™
Lithium carbonate

Powder. For lab use only.

Formula: Li₂CO₃
MW: 73.89 g/mol
Melting Pt: 720
Density: 2.11 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011084
CAS Number: 554-13-2
EINECS: 209-062-5
Merck Index: 13,05549

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (Li₂CO₃) ≥99.0 %
ACS - Calcium (Ca) ≤0.01 %
ACS - Chloride (Cl) ≤0.005 %
ACS - Heavy Metals ≤0.002 %
ACS - Insoluble in Dilute HCl ≤0.01 %
ACS - Iron (Fe) ≤0.002 %
ACS - Nitrate (NO₃) ≤5 ppm
ACS - Potassium (K) ≤0.01 %
ACS - Sodium (Na) ≤0.1 %
ACS - Sulfur Compounds (as SO₄) ≤0.2 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR