Lanthanum(III) oxide ≥98%, BAKER ANALYZED®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC


Management of Change (MOC) category = N

BAKRP351-05 BAKRP351-06
CAJTP351-5EA 423.09 CAD
CAJTP351-5 CAJTP351-6
Lanthanum(III) oxide ≥98%, BAKER ANALYZED®
Lanthanum (III) oxide

Lot analysis on label. For flame enhancement in atomic absorption spectroscopy.

Formula: La₂O₃
Boiling Pt: 4200 °C (1013 hPa)
Melting Pt: 2315 °C
Density: 6.51 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011071
CAS Number: 1312-81-8
EINECS: 215-200-5

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (La₂O₃) ≥98.0 %
Insoluble Matter ≤0.01 %
Calcium (Ca)(by AAS) ≤0.001 %
Trace Impurities - Magnesium (Mg)(by AAS) ≤2.000 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR