Lithium nitrate ≥97.0% (by acidimetry), BAKER ANALYZED®, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Warning
|
Management of Change (MOC) category = N
BAKR2384-01
BAKR2384-05
CAJT2384-1EA
724.87
CAD
CAJT2384-1
CAJT2384-5
Lithium nitrate ≥97.0% (by acidimetry), BAKER ANALYZED®, J.T.Baker®
Lithium nitrate
Crystals.
Formula:
LiNO₃ MW: 68.95 g/mol Melting Pt: 255 Density: 2.38 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011094 CAS Number: 7790-69-4 EINECS: 232-218-9 UN: 2722 ADR: 5.1 III Merck Index: 13,05558 |
Specification Test Results
For Laboratory,Research,or Manufacturing Use | |
Assay (LiNO₃) (by acidimetry) | ≥97.0 % |
Insoluble Matter | ≤0.01 % |
Chloride (Cl) | ≤0.002 % |
Sulfate (SO₄) | ≤0.1 % |
Barium (Ba) | ≤0.002 % |
Heavy Metals (as Pb) | ≤0.001 % |
Trace Impurities - Iron (Fe) | ≤0.001 % |
Calcium (Ca)(by FES) | ≤0.02 % |
Potassium (K), For Information Only | |
Sodium (Na)(by FES) | ≤0.05 % |
Meets Reagent Specifications for testing USP/NF monographs |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...