Lithium chloride 99,0 Argentometric titration, granular, BAKER ANALYZED® ACS

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning


M.O.C. Category N

2370-01 2370-05 2370-07
CAJT2370-7EA 10436.64 CAD
CAJT2370-7 CAJT2370-5 JT2370-1 JT2370-5 JT2370-7
Lithium chloride 99,0 Argentometric titration, granular, BAKER ANALYZED® ACS
Lithium chloride

Granular. Lot analysis on label.

Formula: LiCl
MW: 42.39 g/mol
Boiling Pt: 1382
Melting Pt: 614
Density: 2.07 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011078
CAS Number: 7447-41-8
EINECS: 231-212-3
Merck Index: 13,05550

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (LiCl) (by Ag titrn) ≥99.0 %
ACS - Insoluble Matter ≤0.01 %
ACS - Loss on Drying ≤1.0 %
ACS - Titrable Base (meq/g) ≤0.008
ACS - Sulfate (SO₄) ≤0.01 %
ACS - Barium (Ba) ≤0.002 %
ACS - Heavy Metals (as Pb) ≤0.002 %
ACS - Iron (Fe) ≤0.001 %
ACS - Calcium (Ca) ≤0.01 %
ACS - Potassium (K) ≤0.01 %
ACS - Sodium (Na) ≤0.20 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR