Lanthanum(III) chloride heptahydrate 64,5 - 70,0% Anhydrous basis, BAKER ANALYZED® ACS

Supplier: AVANTOR PERFORMANCE MATERIAL LLC


M.O.C. Category N

BAKR2255-05 BAKR2255-07
CAJT2255-5CS 2045.44 CAD
CAJT2255-5 CAJT2255-07
Lanthanum(III) chloride heptahydrate 64,5 - 70,0% Anhydrous basis, BAKER ANALYZED® ACS
Lanthanum (III) chloride heptahydrate

For flame enhancement in atomic absorption spectroscopy. Lot analysis on label.


For flame enhancement in atomic absorption spectroscopy.

Formula: LaCl₃·7H₂O
MW: 371.37 g/mol
Boiling Pt: 860 °C (1013 hPa)
Melting Pt: 96
Density: 3.8773 g/cm³ (–0.15 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149756
CAS Number: 10025-84-0
EINECS: 233-237-5
Merck Index: 13,05379

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (LaCl₃) 64.5 - 70.0 %
ACS - Insoluble Matter ≤0.01 %
ACS - Calcium (Ca) ≤0.001 %
Trace Impurities - Magnesium (Mg) ≤10 ppm
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR