HyFlex® Foam Gloves, Ansell
Supplier: Ansell Healthcare
205673 205675 205676 1180011060 1180011080 1180011090
89012-342PK 37.59 CAD
89012-342 89012-344 89012-348 89012-350 89012-352 CA33890-042 CA33890-046 CA33890-048
HyFlex® Foam Gloves, Ansell
Gloves
These gloves provide comfort and dexterity when working with small, oily parts.
  • Hand-specific
  • Excellent abrasion resistance
  • Elastic cuffs
The nitrile coating helps reduce hand fatigue and channels oil away from the glove surface, providing a surer grip. Fine-gauge, ultra-lightweight knit liner offers excellent resistance to nicks and abrasions.