Lactic acid 85%, BAKER ANALYZED® ACS

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: DL-Lactic acid, 2-Hydroxypropionic acid, 2-Hydroxypropanoic acid

M.O.C. Category N

BAKR0194-01 0194-03
CAJT0194-1CS 3175.15 CAD
CAJT0194-1 JT0194-3
Lactic acid 85%, BAKER ANALYZED® ACS
Lactic acid

Lot analysis on label.

Formula: H₃CCH(OH)COOH
MW: 90.08 g/mol
Density: 1.20 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00004520
CAS Number: 50-21-5
EINECS: 200-018-0
UN: 3265
ADR: 8 II
Merck Index: 13,05351

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (as CH₃CHOHCOOH) 85.0 - 90.0 %
ACS - Residue after Ignition ≤ 0.02 %
ACS - Chloride (Cl) ≤ 0.001 %
ACS - Sulfate (SO₄) ≤ 0.002 %
ACS - Substances Darkened by H₂SO₄ Passes Test
ACS - Heavy Metals (by ICP-OES) ≤ 5 ppm
Trace Impurities - Iron (Fe) ≤ 5 ppm
The reagent generally available is a mixture of lactic acid (CH₃CHOHCOOH) and lactic acid lactate (C₆H₁₀O₅).


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR