Lithium hydroxide monohydrate ≥98.0%, GR ACS, Supelco®

Supplier: MilliporeSigma
Danger

LX0350-1 LX0350-5
CALX0350-1EA 246 CAD
CALX0350-1 CA-LX0350-5
Lithium hydroxide monohydrate ≥98.0%, GR ACS, Supelco®
Lithium hydroxide monohydrate

Meets ACS Specifications

Formula: LiOH·1H₂O
MW: 41.96 g/mol
Boiling Pt: 920 °C (1013 hPa)
Melting Pt: 462 °C
Density: 1.51 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149772
CAS Number: 1310-66-3
EINECS: 215-183-4
UN: 2680
ADR: 8,II
Merck Index: 13,05556

Order Now

Specification Test Results

Assay 98.0% min
Chloride (Cl) 0.01% max
Color White
Form Crystals
Heavy metals (as Pb) 0.002% max
Insoluble matter 0.01% max
Iron (Fe) 0.002% max
Lithium carbonate (Li2CO3) 2.0% max
Sulfate (SO4) 0.05% max

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR