Lactic acid ≥85% in aqueous solution, GR ACS, Supelco®
Supplier: MilliporeSigma
|
Danger
|
LX0020-3
LX0020-6
CALX0020-3EA
606
CAD
CALX0020-3
CALX0020-6
Lactic acid ≥85% in aqueous solution, GR ACS, Supelco®
Lactic acid
Meets ACS Specifications
Formula:
H₃CCH(OH)COOH Boiling Pt: 122 °C (20 hPa) Density: 1.11…1.21 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00004520 CAS Number: 50-21-5 EINECS: 200-018-0 |
Specification Test Results
Assay | 85.0-90.0% |
Chloride (Cl) | 0.001% max |
Color (APHA) | 50 max |
Form | Liquid |
Heavy metals (as Pb) | 5ppm max |
Iron (Fe) | 5ppm max |
Residue after ignition | 0.02% max |
Substances darkened by sulfuric acid | To pass test |
Sulfate (SO4) | 0.002% max |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...