KONTES® Round-Bottom Boiling Flasks, Vertical Type, Necks, Heavy Wall, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
606000-0224 606000-0624 606000-1024 606000-1424 606000-4824 606020-1024 606020-1324 606020-4829 606000-1129 606000-1524 606000-1529 606000-1729 606000-1924 606000-1929 606000-2224 606020-1124 606020-1129 606020-1424 606020-1824 606020-1924
46200-670EA 405.89 CAD
46200-670 KT606000-0224 KT606000-0624 KT606000-1024 KT606000-1424 KT606000-1824 KT606000-2524 KT606000-2624 KT606000-4824 KT606020-0224 KT606020-0624 KT606020-1024 KT606020-1324 KT606020-2624 KT606020-2924 KT606020-4829 KT606000-1124 KT606000-1129 KT606000-1524 KT606000-1529 KT606000-1729 KT606000-1924 KT606000-1929 KT606000-2124 KT606000-2224 KT606000-2324 KT606000-2329 KT606000-2924 KT606020-1124 KT606020-1129 KT606020-1424 KT606020-1529 KT606020-1824 KT606020-1924 KT606020-2124 KT606020-2224 KT606020-2724
KONTES® Round-Bottom Boiling Flasks, Vertical Type, Necks, Heavy Wall, Kimble Chase
Flasks Round Bottom Flasks
The tops of all three necks are approximately the same height, with Standard Taper outer joints.
  • Side necks are vertical
Designed from ASTM Specification E1378, Type IV requirements. Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Certifications: Designed from ASTM Specification E1378, Type IV requirements.
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Ordering information: For additional volumes and [ST] joints, contact your Avantor sales representative.