Lanthanum(III) oxide ≥99.99% (REO, rare earth oxide basis), REacton®
Supplier: Thermo Scientific Chemicals
|
Warning
|
11264-18
11264-30
11264-A1
CAAA11264-18EA
83.27
CAD
CAAA11264-18
CAAA11264-30
CAAA11264-A1
Lanthanum(III) oxide ≥99.99% (REO, rare earth oxide basis), REacton®
Lanthanum (III) oxide
Powder. Hygroscopic. Purity based on rare earth contaminates.
Purity: 99.99%
CAS No.: 1312-81-8
Molecular Formula: La2O3
Formula Weight: 325.82
Physical Form: Powder
Density: 6.51
Melting Point: 2307°
Boiling Point: 4200°
Merck Reference: 14,5363
MDL No.: MFCD00011071
Formula:
La₂O₃ MW: 325.81 g/mol Boiling Pt: 4200 Melting Pt: 2315 Density: 6.51 Storage Temperature: Ambient |
MDL Number:
MFCD00011071 CAS Number: 1312-81-8 EINECS: 215-200-5 Merck Index: 14,5363 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...