di-Lithium tetraborate 99,998 Metals basis, Puratronic®

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Lithium tetraborate, Dilithium tetraborate

10744-14 10744-22 10744-A1
CAAA10744-14EA 98.43 CAD
CAAA10744-14 CAAA10744-22 CAAA10744-A1
di-Lithium tetraborate 99,998 Metals basis, Puratronic®
di-Lithium tetraborate

Powder, 22 mesh. Purity based on metal contaminates only.


Purity: 99.998% CAS No.: 12007-60-2 Molecular Formula: B4Li2O7 Formula Weight: 169.12 Physical Form: -22 Mesh Powder Density: 2.4 Appearance: White Odor: Odorless Melting Point: 930° Merck Reference: 14,5524 MDL No.: MFCD00011083

Formula: Li₂B₄O₇
MW: 169.12 g/mol
Melting Pt: 918
Density: 2.4
Storage Temperature: Ambient
MDL Number: MFCD00011083
CAS Number: 12007-60-2
EINECS: 234-514-3
Merck Index: 14,05524

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR