KIMAX® Flat-Bottom Boiling Flasks, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
25055-300 25055-250 25055-500 25055-1000
29114-050EA 103.59 CAD
29114-050 89001-264 89001-266 89001-268
KIMAX® Flat-Bottom Boiling Flasks, Kimble Chase
Flasks Round Bottom Flasks
All flasks have a short neck and a full length [ST] 24/40 interchangeable ground glass joint
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.