Kerosene, Supelco®

Supplier: MilliporeSigma
Danger

KX0020-20 KX0020-3 KX0020-1
CAKX0020-20EA 4706 CAD
CAKX0020-20 CAKX0020-3 CAKX0020-1
Kerosene, Supelco®
Kerosene - CAS 64742-48-9

GR

Boiling Pt: 180…188 °C
Storage Temperature: Ambient
CAS Number: 64742-48-9
UN: 1268
ADR: 3,III

Order Now

Specification Test Results

Boiling range 171-191°C
Color (APHA) 20 max
Form Clear odorless liquid
Infrared spectrum Conforms to standard
Specific Gravity at 15.6/15.6°C 0.756-0.761
Sulfur (S) 1ppm max

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR