Digital Brix/Refractive Index Refractometers
Supplier: Heathrow Scientific
48101 48102
76545-880EA 593.91 CAD
76545-880 76545-882
Digital Brix/Refractive Index Refractometers
Refractometers Handheld Refractometers
The MDX-series delivers digital accuracy and high-end features for specific applications. Brix and Refractive Index scales can be used in a wide variety of industries including: food and beverage, industrial, waste-water, aquatics, and many more.
  • Compact and lightweight
  • Removable grip and prism cover for easy cleaning
  • Continuous loop read features (15 reads)
  • Automatic Temperature Control
  • Zero with distilled water
  • 589.3 nm LED
  • Stainless steel prism well
  • °C/°F switchable temperature display
  • 1 year warranty
The MDX-series Digital Brix/SugarMore Product Information