4-Isopropoxyaniline ≥98.0% (by GC, titration analysis)

Supplier: TCI America

I0399-100G I0399-5G
TCI0399-100GEA 511.92 CAD
TCI0399-100G TCI0399-5G
4-Isopropoxyaniline ≥98.0% (by GC, titration analysis)
4-Isopropoxyaniline

CAS Number: 7664-66-6
MDL Number: MFCD00059188
Molecular Formula: C9H13NO
Molecular Weight: 151.21
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 95
Specific Gravity (20/20): 1.03

Formula: C₉H₁₃NO
Boiling Pt: 95 °C
Density: 1.03 (20 °C)
MDL Number: MFCD00059188
CAS Number: 7664-66-6
EINECS: 231-636-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3dc4ecf59627cba8d21a59f232cf0935086f15ef&vid=3dc4ecf59627cba8d21a59f232cf0935086f15ef&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR