(±)-Isonipecotic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

I0256-25G I0256-500G I0256-100G
TCI0256-025GEA 32.15 CAD
TCI0256-025G TCI0256-500G TCI0256-100G
(±)-Isonipecotic acid ≥98.0% (by titrimetric analysis)
(±)-Isonipecotic acid

CAS Number: 498-94-2
MDL Number: MFCD00006004
Molecular Formula: C6H11NO2
Molecular Weight: 129.16
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 336


Product Categories: Bioscience, non-Proteinogenic Amino Acids
Other Product Names / Synonyms: Isonipecotic Acid

Formula: C₆H₁₁NO₂
Boiling Pt: 264 °C (1013 hPa)
Melting Pt: 336 °C
MDL Number: MFCD00006004
CAS Number: 498-94-2
EINECS: 207-872-3
Merck Index: 13,05208
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bdbf5bc64a5f73f64dc73848fd00478c1e257379&vid=bdbf5bc64a5f73f64dc73848fd00478c1e257379&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR