Isonicotinic acid ≥99.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

I0207-25G I0207-500G
TCI0207-025GEA 36.99 CAD
TCI0207-025G TCI0207-500G
Isonicotinic acid ≥99.0% (by HPLC, titration analysis)
Isonicotinic acid

CAS Number: 55-22-1
MDL Number: MFCD00006429
Molecular Formula: C6H5NO2
Molecular Weight: 123.11
Purity/Analysis Method: >99.0% (HPLC,T)
Form: Crystal
Melting point (°C): 315


Product Categories: Materials Chemistry, Metal Organic Framework (MOF) and Porous Coordination Polymer (PCP) Research Reagents
Other Product Names / Synonyms: Pyridine-4-carboxylic Acid

Formula: C₆H₅NO₂
Boiling Pt: 396 °C (1013 hPa)
Melting Pt: 315 °C
Storage Temperature: Ambient
MDL Number: MFCD00006429
CAS Number: 55-22-1
EINECS: 200-228-2
Merck Index: 13,05206
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9d3a1153ec3f01fec0c4c009a47f21ccd7f26c28&vid=9d3a1153ec3f01fec0c4c009a47f21ccd7f26c28&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR