Dimethyl itaconate ≥98.0% stabilized
Supplier: TCI America
I0206-25G
I0206-500G
TCI0206-025GEA
49.31
CAD
TCI0206-025G
TCI0206-500G
Dimethyl itaconate ≥98.0% stabilized
Dimethyl itaconate
(stabilized with HQ)
CAS Number: 617-52-7
MDL Number: MFCD00008446
Molecular Formula: C7H10O4
Molecular Weight: 158.15
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 109
Flash Point (°C): 114
Freezing point (°C): 37
Product Categories: Materials Chemistry, Copolymerization Monomers
Other Product Names / Synonyms: Itaconic Acid Dimethyl Ester
Formula:
C₇H₁₀O₄ Boiling Pt: 109 °C Melting Pt: 38 °C Density: 1.1399 g/cm³ (14.7 °C) Flash Pt: 114 °C |
MDL Number:
MFCD00008446 CAS Number: 617-52-7 EINECS: 210-519-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...