Ionic strength adjustor (ISA) for use with sodium ion selective electrode
Supplier: Ricca Chemical
R4130000-500A
R4130000-1A
RC413016EA
86.9
CAD
RC413016
RC413032
Ionic strength adjustor (ISA) for use with sodium ion selective electrode
Ionic strength adjustor (ISA)
For use with Sodium Ion Selective Electrode.
Specification Test Results
Appearance | Clear, colorless, ammonia odor |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...