KIMAX® Flat-Bottom Florence Boiling Flasks, Reinforced Tooled Top, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
25000-500 25000-1000 25000-6000
89000-980CS 273.35 CAD
89000-980 89000-982 89001-260
KIMAX® Flat-Bottom Florence Boiling Flasks, Reinforced Tooled Top, Kimble Chase
Flasks Round Bottom Flasks
Flasks have a low coefficient of expansion to resist thermal shock
They are constructed with sturdy walls to minimize mechanical breakage and with reinforced tooled tops for strength and a secure stopper fit.
Order Now
SPECIFICATIONS
Environmentally Preferable