KIMAX® Round-Bottom Boiling Flasks, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
25285-1000 25285-50 25285-100 25285-500 25285-2000
89000-778CS 1289.28 CAD
89000-778 89001-280 89001-282 89001-290 89001-292 89001-294
KIMAX® Round-Bottom Boiling Flasks, Kimble Chase
Flasks Round Bottom Flasks
Short neck flasks are designed to ASTM Specification E1403, Class 3 requirements
They feature a full-length, outer, [ST] 24/40 joint.