Iodine monochloride solution_SC acc. Wijs USP Test Solution (TS) Iodine Absorption Number of Fat and Oils
Supplier: Ricca Chemical
|
Danger
|
Synonyms:
Iodochloride
4110-1
R4110000-500C
4110-32
4110-5PT
RC41101EA
1136.02
CAD
RC41101
RC411016
RC411032
RC41105PT
Iodine monochloride solution_SC acc. Wijs USP Test Solution (TS) Iodine Absorption Number of Fat and Oils
Iodine monochloride
Meets AOAC standards for iodine absorption number.
Formula:
ClI MW: 162.36 g/mol Boiling Pt: 97.3 Melting Pt: 26 Flash Pt: 39 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011354 CAS Number: 7790-99-0 EINECS: 232-236-7 UN: 2920 ADR: 8 (3) II Merck Index: 13,05039 |
Specification Test Results
Appearance | Clear, dark brown, Acetic/Halogen odor |
Iodine/Chlorine Ratio | 1.10 ± 0.05 Iodine/Chlorine Ratio |
Total Halogen Assay at 20 °C (traceable to NIST SRM 136) | 0.191 ± 0.002 N at 20.0 °C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...