Iodine 0.01 N (N/100) in aqueous solution, StandARd® volumetric solution, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC


Management of Change (MOC) category = N

H175-05 MACRH175-07
MKH17505CS 588.47 CAD
MKH17505 CAMKH17507
Iodine 0.01 N (N/100) in aqueous solution, StandARd® volumetric solution, Macron Fine Chemicals™
Iodine

Contains 0.1–0.4% iodine and Max. 1% potassium iodide.


Contains 0.1–0.4% iodine and Max. 1% potassium iodide.

Formula: I₂
MDL Number: MFCD00011355
CAS Number: 7553-56-2
ADR: 8,III

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Normality 0.0095 - 0.0105
Appearance (transparent, reddish-brown solution)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR