Iron(III) sulfate hydrate ≥73.0%, granular, AR®, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning


Management of Change (MOC) category = N

MACR5036-05 MACR5036-12
CAMK503605EA 4263.39 CAD
CAMK503605 CAMK503612
Iron(III) sulfate hydrate ≥73.0%, granular, AR®, Macron Fine Chemicals™
Iron (III) sulphate hydrate

Powder.

Formula: Fe₂(SO₄)₃·xH₂O
Density: 3.1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149714
CAS Number: 15244-10-7
EINECS: 233-072-9
Merck Index: 13,04065

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (Fe₂(SO₄)₃) ≥73.0 %
Solubility Passes Test
Insoluble Matter ≤0.020 %
Chloride (Cl) ≤0.005 %
Ferrous Iron ≤0.02 %
Nitrate (NO₃) ≤0.01 %
Substances not Precipitated by NH₄OH ≤0.10 %
Copper (Cu) ≤0.005 %
Zinc (Zn) ≤0.005 %
Appearance (tan to light greenish-yellow. Granular or granular powder.)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR