Precision Specific Gravity Hydrometers
Supplier: Heathrow Scientific
6603-10 6603-11 6603-12 6603-13 6603-2 6603-21 6603-22 6603-3 6603-4 6603-5 6603-6 6603-7 6603-8 6603-9
76546-374EA 44.57 CAD
76546-374 76546-376 76546-378 76546-380 76546-382 76546-384 76546-386 76546-388 76546-390 76546-392 76546-394 76546-396 76546-398 76546-400
Precision Specific Gravity Hydrometers
Hydrometers
These precision specific gravity hydrometers measure the ratio of the mass of a solid or liquid to the mass of an equal volume of distilled water.
Each hydrometer has been calibrated for use at 60 °F.
Order Now
SPECIFICATIONS
Environmentally Preferable