Ultra Precision Specific Gravity Hydrometers
Supplier: Heathrow Scientific
6602-10 6602-11 6602-12 6602-13 6602-14 6602-15 6602-19 6602-2 6602-3 6602-4 6602-5 6602-6 6602-7 6602-8 6602-9
76546-324EA 47.73 CAD
76546-324 76546-328 76546-330 76546-332 76546-334 76546-336 76546-338 76546-340 76546-342 76546-344 76546-348 76546-352 76546-356 76546-360 76546-364
Ultra Precision Specific Gravity Hydrometers
Hydrometers
These ultra precision specific gravity hydrometers measure the ratio of the mass of a solid or liquid to the mass of an equal volume of distilled water.
These hydrometers feature smaller ranges but with higher resolution and have all been calibrated for use at 60 °F.