Baume Hydrometers
Supplier: Heathrow Scientific
6609-1 6609-10 6609-11 6609-12 6609-13 6609-14 6609-15 6609-2 6609-3 6609-4 6609-5 6609-8 6609-9
76546-446EA 44.57 CAD
76546-446 76546-448 76546-450 76546-452 76546-454 76546-456 76546-458 76546-460 76546-462 76546-464 76546-466 76546-468 76546-470
Baume Hydrometers
Hydrometers
VEE GEE scientific Baume scale hydrometers are for liquids heavier than water. The Baume scale is based on the following: Specific gravity = 145/(145 - degrees Baume).
  • Wide variety of Baume ranges available
Calibrated for use at 15.5 °C/60 °F.