Plato Hydrometers with Thermometer
Supplier: Heathrow Scientific
6614TS-5 6614TS-6 6614TS-7 6614TS-8
76546-530EA 86.82 CAD
76546-530 76546-532 76546-534 76546-536
Plato Hydrometers with Thermometer
Hydrometers
Our Plato hydrometers are calibrated to show the concentration of extract as a percentage by weight in brewery wort. For example, a 5° P reading indicates 5 g of extract per 100 g of wort.
  • Calibration for use at 20 °C/68 °F
  • Spirit thermometer liquid fill
Primarily used in the brewing industry, the Plato scale is carried out at 20 °C.