Brix Hydrometers with Thermometer (°F)
Supplier: Heathrow Scientific
6601TS-10F 6601TS-13F 6601TS-1F 6601TS-2F 6601TS-3F
76546-294EA 85.58 CAD
76546-294 76546-300 76546-306 76546-310 76546-314
Brix Hydrometers with Thermometer (°F)
Hydrometers
Our brix scale hydrometers with ℉ thermometer are calibrated to show the percentage of sucrose by weight at 68 ℉.
  • Calibrated to show percent sucrose by weight
  • Wide variety of brix ranges available
  • Thermometer liquid fill is safety blue
The Brix scale is based on the following: 1° Brix = 1% sucrose by weight at specified temperature (68 ℉).
The thermometer scale is located in the body and contains safety blue liquid fill. The thermometers have a range of 30 to 150 ℉ with 2° subdivisions. A correction scale for conversion of reading to 68 ℉ is printed opposite the temperature scale.