Imidazole ≥99.0%, OR, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Synonyms:
Glyoxaline
, 1H-Imidazole
, Glyoxalamine
Management of Change (MOC) category = NI
MACR4337-55
MACR4337-09
4337-55
4337-09
CAMK433709EA
2103.69
CAD
MK433755
CAMK433709
CAMK433755
MK433709
Imidazole ≥99.0%, OR, Macron Fine Chemicals™
Imidazole
Macron Fine Chemicals™ OR: organic reagents that are suitable for research applications.
Formula:
C₃H₄N₂ MW: 68.08 g/mol Boiling Pt: 255 °C (1013 hPa) Melting Pt: 89…90 °C Density: 1.18 g/cm³ (20 °C) Flash Pt: 135 °C |
MDL Number:
MFCD00005183 CAS Number: 288-32-4 UN: 3263 ADR: 8,III Merck Index: 13,04935 |
Specification Test Results
For Laboratory,Research,or Manufacturing Use | |
Assay | ≥99.0 % |
Water (H₂O) | ≤0.5 % |
Appearance (white to off white flakes or crystalline powder) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...