Imidazole ≥99.0%, BAKER ANALYZED® ACS
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Danger
|
Synonyms:
Glyoxaline, 1H-Imidazole, Glyoxalamine
Management of Change (MOC) category = NI
BAKRN811-05
CAJTN811-6
BAKRN811-07
CAJTN811-5EA
245.37
CAD
CAJTN811-5
CAJTN811-6
CAJTN811-07
Imidazole ≥99.0%, BAKER ANALYZED® ACS
Imidazole
Formula:
C₃H₄N₂ MW: 68.08 g/mol Boiling Pt: 255 Melting Pt: 89…90 °C Density: 1.18 g/cm³ (20 °C) Flash Pt: 135 Storage Temperature: Ambient |
MDL Number:
MFCD00005183 CAS Number: 288-32-4 EINECS: 206-019-2 UN: 3263 ADR: 8 III Merck Index: 13,04935 |
|
Specification Test Results
Meets ACS Reagent Chemical Requirements | |
For Laboratory,Research,or Manufacturing Use | |
Assay (C₃H₄N₂) | ≥99.0 % |
Trace Impurities - Iron (Fe) | ≤0.001 % |
pH of 5% Solution at 25°C | 9.5 - 11.0 |
Residue after Ignition | ≤0.10 % |
Water (H₂O) | ≤0.2 % |
DNase Activity, For Information Only | Passes Test |
Protease Activity, For Information Only | Passes Test |
RNase Activity, For Information Only | Passes Test |
Meets Reagent Specifications for testing USP/NF monographs |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...