KIMTECH PURE® G5 STERLING™ Nitrile Gloves, Kimberly-Clark
Supplier: Avantor
CA89093-004CS 588.63 CAD
CA89093-004 CA89093-006 CA89093-008 CA89093-010 CA89093-012
KIMTECH PURE® G5 STERLING™ Nitrile Gloves, Kimberly-Clark
Gloves
These gloves provide the protection of nitrile and sensitivity comparable to natural rubber latex.
  • Powder-free, latex-free, silicone-free
  • Tactile sensitivity with tensile strength
  • Static dissipative in use
  • Ambidextrous, beaded cuff, textured fingertips
Recommended for ISO Class 5 (FED-STD-209E Class 100/M3.5) or higher cleanroom environments. Suitable for use in the manufacture of pharmaceuticals, medical devices, electronics, disk drives, flat panel displays, and semiconductors. Packaged in a Class 100 cleanroom.
Certifications: AQL 1.5.