PYREX® Boiling Flasks, Flat Bottom, Short Neck, Standard Taper Joint, Corning
Supplier: Corning
PYREX®
4100-250 4100-125 4100-1L 4100-500 4100-50
89090-796CS 914.62 CAD
89090-796 29113-060 89091-136 89090-692 89090-882
PYREX® Boiling Flasks, Flat Bottom, Short Neck, Standard Taper Joint, Corning
Flasks Flat Bottom Flasks
PYREX® borosilicate glass, clear, with short neck.
  • High thermal shock resistance
  • Chemically inert
  • Provide contamination free results
  • Long life
These flasks have full length outer standard tapered 24/40 joint. The normal working temperature is 230 °C (446 °F).