Plastic Bracket for Hard Hats, Honeywell Safety
Supplier: Honeywell Safety
CP5006 CP5005 CP5005 CP5006
CA13502-278EA 27.77 CAD
CA13502-278 13502-278 13502-281 13502-280
Plastic Bracket for Hard Hats, Honeywell Safety
Head Protection Safety Helmets
Lightweight plastic bracket fits into any slotted hard hat
Bracket 13502-278 fits Everest wide brim hats only (see 13502-144 series).
Bracket accommodates ORIS® earmuff systems. Bracket cam lock system accepts a wide range of face shields.