Ammonium iron (II) sulfate hexahydrate 98.5-101.5% (by KMnO₄ titration), fine crystals, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning


Management of Change (MOC) category = N

BAKR2054-01 CAJT2054-19 BAKR2054-05
CAJT2054-7EA 5762.37 CAD
CAJT2054-7 CAJT2054-1 CAJT2054-19 CAJT2054-5 CAJT2054-69
Ammonium iron (II) sulfate hexahydrate 98.5-101.5% (by KMnO₄ titration), fine crystals, BAKER ANALYZED® ACS, J.T.Baker®
Ammonium iron (II) sulphate hexahydrate

Fine crystals. Lot analysis on label.


Caution: Due to inherent oxidation, ferric ion (Fe³⁺) content may increase on storage.

Formula: (NH₄)₂Fe(SO₄)₂·6H₂O
Melting Pt: 100 °C
Density: 1.864 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00150530
CAS Number: 7783-85-9
EINECS: 233-151-8

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (Fe(NH₄)₂(SO₄)₂·6H₂O) (by KMnO₄ titrn) 98.5 - 101.5 %
Insoluble Matter ≤0.01 %
pH of 5% Solution at 25°C 3.0 - 5.0
Chloride (Cl) ≤0.001 %
Phosphate (PO₄) ≤0.003 %
Calcium (Ca) ≤0.005 %
Copper (Cu) ≤0.003 %
Ferric Ion (Fe³⁺) ≤0.01 %
ACS - Magnesium (Mg) ≤0.002 %
Manganese (Mn) ≤0.01 %
Potassium (K) ≤0.002 %
Sodium (Na) ≤0.02 %
Zinc (Zn) ≤0.003 %
NOTE: Due to inherent oxidation, ferric ion (Fe³⁺) content may increase
on storage.
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR