Wide Brim Hard Hats, Honeywell Safety
Supplier: Honeywell Safety
A49070000
CA13502-150CS 385.78 CAD
CA13502-150
Wide Brim Hard Hats, Honeywell Safety
Head Protection Safety Helmets
The Everest style high-density polyethylene hard hats feature a wide brim for added protection against UV rays, rain, and falling debris
  • High-density polyethylene protection against UV rays, rain, and falling debris
  • Accessory slots for face and hearing protection accessories
  • Suspension types offer forehead comfort band and crown pad, two-level adjustment, and self-centering
Accessory slots allow attachment of face and hearing protection accessories. Hats are available with four-point suspension with pin lock adjustment, or with six-point suspension with ratchet adjustment. Both suspension types offer a forehead comfort band and crown pad, two-level adjustment, and self-centering.
Certifications: CE marked. Meet ANSI Type I Class E requirements.