KIMBLE® Recovery Flasks, Heavy Wall with Standard Joint Bead, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
294300-0025 294300-0050 294300-0100 294300-0200 294305-0025 294305-0050
KT294300-0010EA 94.67 CAD
KT294300-0010 KT294300-0025 KT294300-0050 KT294300-0100 KT294300-0200 KT294305-0025 KT294305-0050
KIMBLE® Recovery Flasks, Heavy Wall with Standard Joint Bead, DWK Life Sciences
Flasks Rotary Evaporator Flasks
Single neck flask with a Standard Taper outer joint designed for easy recovery of reaction products.
  • Total contents are accessible with a spatula or policeman
  • Lower portion fits standard heating mantles
  • Manufactured from 33 expansion, low extractable borosilicate glass
Even wall thickness and minimum runout make these flasks ideal for use with rotary evaporators.
Certifications: Borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.