KONTES® AIRLESS-WARE® Flasks, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
213100-0214 213100-0514 213100-1024 213100-2014 213100-2024 213100-5024 213100-1014
KT213100-0214EA 323.41 CAD
KT213100-0214 KT213100-0514 KT213100-1024 KT213100-2014 KT213100-2024 KT213100-5024 KT213100-0114 KT213100-1014
KONTES® AIRLESS-WARE® Flasks, Kimble Chase
Flasks Kjeldahl Flasks
Improved version of classic Schlenk tube
The modified Kjeldahl shape facilitates heating, stirring and solvent stripping, without impairing ease of transfer of liquids and solids. Bottom of flask shaped to fit KONTES® spherical heating mantles. Supplied with KT801001-0002 glass plug and KT809000-0021 pressure/vacuum retainer.