KIMAX® Kjeldahl Flasks, Kimble Chase
Supplier: DWK LIFE SCIENCES S. DE R.L. DE MX
27400-100 27400-500 27400-800
89001-842CS 1229.28 CAD
89001-842 89001-844 89001-848 89001-850
KIMAX® Kjeldahl Flasks, Kimble Chase
Flasks Kjeldahl Flasks
All flasks have reinforcing beads at the top
The 500mL and 800mL flasks have tooled necks to provide for accurate fit of stoppers and to reduce possibility of breakage. The 30mL size is designed from ASTM Specification E147. The 100mL and larger sizes are designed from ASTM Specification E1377, Type I requirements.