Tranquility® Nitrile Gloves, Microflex®, Ansell
Supplier: Ansell
Tranquility®
TQ-601-XS TQ-601-S TQ-601-M TQ-601-XL TQ-601-XL TQ-601-S TQ-601-XS TQ-601-M TQ-601-L TQ-601-L
94024-064CS 440.91 CAD
94024-064 CA94024-062 94024-056 CA94024-064 CA94024-056 94024-058 CA94024-058 94024-060 CA94024-060 94024-062
Tranquility® Nitrile Gloves, Microflex®, Ansell
Gloves
Latex-free nitrile gloves contain no natural rubber proteins and are soft, thin, and conform to the hand for excellent dexterity and grip. These gloves now feature HYDRASOFT™ Skin Moisturizing Technology.
  • Ambidextrous
  • Beaded cuffs
  • Powder-free
  • Textured fingers
Recommended when enhanced comfort and tactile sensitivity are a must for everyday use of small instruments. The HYDRASOFT™ Skin Moisturizing Technology is proven to allow skin to retain 2X more moisture.
Certifications: Gloves meet or exceed ASTM D6319 for physical properties.