Iron(II) sulfate heptahydrate 99.5-102.0% (by manganometry), EMSURE® ISO, ACS, Reag. Ph. Eur. analytical reagent, Supelco®

Supplier: MilliporeSigma
Warning

Synonyms: Iron vitriol, Ferrous sulphate heptahydrate, Ferrous sulfate heptahydrate

M-Clarity™ quality level = MQ300

1.03965.1000 1.03965.5000 1.03965.0500 1.03965.0100 1.03965.9025
CA1.03965.1000EA 335 CAD
CA1.03965.1000 CA1.03965.5000 CA1.03965.0500 CA1.03965.0100 CA1.03965.9025
Iron(II) sulfate heptahydrate 99.5-102.0% (by manganometry), EMSURE® ISO, ACS, Reag. Ph. Eur. analytical reagent, Supelco®
Iron (II) sulphate heptahydrate

EMSURE® (formerly GR) is a premium grade of reagents for wet chemical analysis, adhering to ACS, ISO, and/or Reag. Ph.Eur quality specifications.


1.03965.0500 replaces and is the same as FX0260-1. Part # Change only, Same Pack Size, Same Specifications. 1.03965.1000 is offered as an alternative to FX0260-5. Different Pack Size, Same Specifications.

Formula: FeSO₄·7H₂O
MW: 278.02 g/mol
Melting Pt: 64 (–H₂O of crystall.)
Density: 1.89 g/cm3 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149719
CAS Number: 7782-63-0
EINECS: 231-753-5
Merck Index: 13,04091

Order Now

Specification Test Results

Assay (manganometric) 99.5 - 102.0 %
Identity passes test
Appearance of solution passes test
Insoluble matter ≤ 0.01 %
pH-value (5 %; water) 3.0 - 4.0
Chloride (Cl) ≤ 0.0005 %
Phosphate (PO₄) ≤ 0.001 %
Total nitrogen (N) ≤ 0.001 %
Heavy metals as Pb ≤ 0.005 %
As (Arsenic) ≤ 0.0002 %
Ca (Calcium) ≤ 0.005 %
Cr (Chromium) ≤ 0.0050 %
Cu (Copper) ≤ 0.001 %
Fe III (Iron III) ≤ 0.02 %
K (Potassium) ≤ 0.002 %
Mg (Magnesium) ≤ 0.002 %
Mn (Manganese) ≤ 0.05 %
Na (Sodium) ≤ 0.02 %
Ni (Nickel) ≤ 0.0050 %
Pb (Lead) ≤ 0.0005 %
Zn (Zinc) ≤ 0.0050 %
Co (Cobalt) ≤ 0.0025 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR