KONTES® Safety-Coated Filtering Flasks, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
953760-0122 953760-1002 953760-2002 953760-4002
KT953760-0122EA 222.47 CAD
KT953760-0122 KT953760-1002 KT953760-2002 KT953760-4002
KONTES® Safety-Coated Filtering Flasks, Kimble Chase
Flasks Filter Flasks
Flasks are manufactured from heavy-wall borosilicate glass and bonded to a plastic safety coating for extra safety during vacuum filtration
The safety coating is autoclavable to 121°C (275°F). All flasks have #2 side arm for connection to 6.4mm (1/4") I.D. flexible tubing.