IPTG (isopropyl-β-D-thiogalactopyranoside) ≥99.0%, OmniPur®

Supplier: MilliporeSigma

5800-1GM 5810-5GM 5815-25GM 5820-100GM
CA-EM5800EA 196.81 CAD
CA-EM5800 CA-EM5810 CA5815 CA5820
IPTG (isopropyl-β-D-thiogalactopyranoside) ≥99.0%, OmniPur®
IPTG (Isopropyl-β-D-thiogalactopyranoside)

White crystalline powder. Inducer for [beta]-galactosidase. Use 1 part IPTG with 4 parts X-Gal. Toxic effects.

Formula: C₉H₁₈O₅S
Boiling Pt: 402 °C (1013 hPa)
Melting Pt: 109.5…110.5 °C
Storage Temperature: Freezer
MDL Number: MFCD00063273
CAS Number: 367-93-1
EINECS: 206-703-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR