Imidazole, OmniPur®, Millipore®

Supplier: MilliporeSigma
Danger

5710-50GM 5720-500GM
CA-EM5710EA 344.26 CAD
CA-EM5710 CA-EM5720
Imidazole, OmniPur®, Millipore®
Imidazole

For molecular biology. DNase- and RNase-free.

Formula: C₃H₄N₂
Boiling Pt: 255 °C (1013 hPa)
Melting Pt: 89…90 °C
Density: 1.18 g/cm³ (20 °C)
Flash Pt: 135 °C
Storage Temperature: Ambient
MDL Number: MFCD00005183
CAS Number: 288-32-4
EINECS: 206-019-2
UN: 2923
ADR: 8,III
Merck Index: 13,04935
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=48eba49e61201d6fffa2bef0c54633a20660c7ef&vid=e7f154e4976b52732539e49bbff60454d5eb82ff&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR